missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 263790050

Catalog No. AC263790050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Potassium phosphate, tribasic monohydrate
27176-10-9
96%
K3PO4·H2O
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
tripotassium;phosphate;hydrate
22031701
96%
13% max. (800°C)
2.564
c.p.
5kg
K3O4P·H2O
MFCD00150410
Solubility in water: soluble. Other solubilities: insoluble in alcohol
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
230.28
230.28
Extra Pure
Plastic Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only