Learn More
Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals
$73.50 - $164.99
Chemical Identifiers
CAS | 27176-10-9 |
---|---|
Molecular Formula | K3O4P·H2O |
Molecular Weight (g/mol) | 230.28 |
MDL Number | MFCD00150410 |
InChI Key | RMNIZOOYFMNEJJ-UHFFFAOYSA-K |
Synonym | tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 |
PubChem CID | 22031701 |
IUPAC Name | tripotassium;phosphate;hydrate |
SMILES | O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC263790250
|
Thermo Scientific Chemicals
263790250 |
25 g | Glass bottle |
Each for $73.50
|
|
||||
AC263795000
|
Thermo Scientific Chemicals
263795000 |
500 g | Glass bottle |
Each for $99.17
|
|
||||
AC263790010
|
Thermo Scientific Chemicals
263790010 |
1 kg | Glass bottle |
Each for $164.99
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27176-10-9 | |
230.28 | |
RMNIZOOYFMNEJJ-UHFFFAOYSA-K | |
22031701 | |
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
K3O4P·H2O | |
MFCD00150410 | |
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 | |
tripotassium;phosphate;hydrate |
Specifications
1340.0°C | |
White | |
25 g | |
K3O4P·H2O | |
MFCD00150410 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] | |
230.28 | |
230.28 | |
Extra Pure | |
Glass bottle | |
Potassium phosphate, tribasic monohydrate, c.p. |
27176-10-9 | |
Powder | |
96% | |
K3PO4·H2O | |
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 | |
RMNIZOOYFMNEJJ-UHFFFAOYSA-K | |
tripotassium;phosphate;hydrate | |
22031701 | |
96% | |
13% max. (800°C) | |
2.564 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis
GHS Signal Word: Danger
RUO – Research Use Only