missing translation for 'onlineSavingsMsg'
Learn More

Potassium Propionate 98.0+%, TCI America™

$61.51 - $404.51

Chemical Identifiers

CAS 327-62-8
Molecular Formula C3H5KO2
Molecular Weight (g/mol) 112.169
MDL Number MFCD00045936
InChI Key BWILYWWHXDGKQA-UHFFFAOYSA-M
Synonym Propionic Acid Potassium Salt
PubChem CID 23663619
IUPAC Name potassium;propanoate
SMILES CCC(=O)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
P051025G
View Documents
TCI America
P051025G
25 g
Each for $61.51
Only null left
 
P0510500G
View Documents
TCI America
P0510500G
500 g
Each for $404.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

327-62-8
112.169
BWILYWWHXDGKQA-UHFFFAOYSA-M
23663619
CCC(=O)[O-].[K+]
C3H5KO2
MFCD00045936
Propionic Acid Potassium Salt
potassium;propanoate

Specifications

327-62-8
C3H5KO2
25 g
BWILYWWHXDGKQA-UHFFFAOYSA-M
potassium;propanoate
23663619
≥98.0% (T)
Potassium Propionate
White
MFCD00045936
Propionic Acid Potassium Salt
CCC(=O)[O-].[K+]
112.169
112.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4132

TSCA : No

SDS
Documents

Documents