Learn More
Potassium sodium L-tartrate tetrahydrate, 99%, Thermo Scientific Chemicals
$98.00 - $1215.89
Chemical Identifiers
CAS | 6381-59-5 |
---|---|
Molecular Formula | C4H12KNaO10 |
Molecular Weight (g/mol) | 282.218 |
MDL Number | MFCD00150989 |
InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
PubChem CID | 2724148 |
IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1016330
|
Thermo Scientific Chemicals
A1016330 |
250 g |
Each for $98.00
|
|
|||||
AAA101630B
|
Thermo Scientific Chemicals
A101630B |
1000 g |
Each for $294.96
|
|
|||||
AAA101630I
|
Thermo Scientific Chemicals
A101630I |
5000 g |
Each for $1,215.89
|
|
|||||
Description
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehilingâs solution and Nylanderâs test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBaking powder, medicine (cathartic), component of Fehling′s solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehilingâs solution and Nylanderâs test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.
Solubility
Soluble in water. Insoluble in alcohol
Notes
Loses water of crystallization at 140°. Unstable above 225°. Incompatible with strong oxidizing agents.
Chemical Identifiers
6381-59-5 | |
282.218 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
2724148 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
C4H12KNaO10 | |
MFCD00150989 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
Specifications
6381-59-5 | |
1.77 | |
C4H12KNaO10 | |
250 g | |
14,7670 | |
Soluble in water; Insoluble in alcohol | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
282.218 | |
282.23 (210.29 Anhydrous) | |
Potassium sodium L-tartrate tetrahydrate |
70°C to 80°C | |
99% | |
MFCD00150989 | |
6113568 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
2724148 | |
99% |
Safety and Handling
EINECSNumber : 206-156-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only