missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Sodium Tartrate Tetrahydrate, 99.0-102.0%, (ACS Reagent Grade), MP Biomedicals
$103.88 - $936.50
Chemical Identifiers
CAS | 6381-59-5 |
---|---|
Molecular Formula | C4H12KNaO10 |
Molecular Weight (g/mol) | 282.218 |
InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
PubChem CID | 2724148 |
IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15256380
|
MP Biomedicals Inc
0215256380 |
100 g |
N/A
|
|
|||||
ICN15256390
|
MP Biomedicals Inc
0215256390 |
500 g |
N/A
|
|
|||||
ICN15256392
|
MP Biomedicals Inc
0215256392 |
3 kg |
N/A
|
|
|||||
Chemical Identifiers
6381-59-5 | |
282.218 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
C4H12KNaO10 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
2724148 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Specifications
6381-59-5 | |
99.0-102.0% | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
282.218 | |
282.22 | |
Potassium Sodium Tartrate Tetrahydrate |
100 g | |
C4H12KNaO10 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
2724148 | |
ACS Reagent |
Safety and Handling
Recommended Storage : Room Temperature