missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals
$136.87 - $775.54
Chemical Identifiers
CAS | 6381-59-5 |
---|---|
MDL Number | MFCD00150989 |
InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
PubChem CID | 2724148 |
IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC202865000
|
Thermo Scientific Chemicals
202865000 |
500 g | Plastic bottle |
Each for $136.87
|
|
||||
AC202860010
|
Thermo Scientific Chemicals
202860010 |
1 kg | Plastic bottle |
Each for $241.17
Case of 6 Each for $0.00
|
|
||||
AC202860051
|
Thermo Scientific Chemicals
202860051 |
5 kg | Plastic Drum |
Each for $775.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6381-59-5 | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
2724148 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
MFCD00150989 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
Specifications
6381-59-5 | |
White | |
Authentic | |
Plastic bottle | |
KO2CCH(OH)CH(OH)CO2Na·4H2O | |
500 g | |
15, 7790 | |
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
282.218 | |
282.22 | |
Analysis | |
Potassium sodium tartrate tetrahydrate |
>100°C | |
5.5 to 8.5 (5% soln. at 20°C) | |
99+% | |
C4H12KNaO10 | |
MFCD00150989 | |
03, 525 | |
+ 22.10 (20.00°C c=10,water) | |
VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
2724148 | |
99 to 102% | |
Crystals |
RUO – Research Use Only