missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Sulfate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical P30410

 View more versions of this product

Catalog No. P304-10


Explore available promotions

Description

Description

Specifications

Specifications

1689°C
1,067°C
White
Solid
≥99 %
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
24507
174.26
Certified ACS
Poly Pail
Potassium Sulfate
7778-80-5
6 to 8
10 kg
K2O4S
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
[K+].[K+].[O-]S([O-])(=O)=O
174.25
CHEBI:32036
≥99%
Pass Test

Chemical Identifiers

7778-80-5
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
dipotassium sulfate
K2O4S
MFCD00011388
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions