missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sulfite, 90%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 440210025

 View more versions of this product

Catalog No. AC440210025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium sulfite
2.5 kg
K2O3S
15,7795
BHZRJJOHZFYXTO-UHFFFAOYSA-L
dipotassium;sulfite
24958
90%
Glass Bottle
10117-38-1
88% min. (Iodimetry)
K2SO3
potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
[O-]S(=O)[O-].[K+].[K+]
158.26
158.26
Pure

Chemical Identifiers

10117-38-1
158.26
potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
dipotassium;sulfite
K2O3S
BHZRJJOHZFYXTO-UHFFFAOYSA-L
24958
[O-]S(=O)[O-].[K+].[K+]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-321-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only