missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™, Thermo Scientific Chemicals

$153.93 - $693.89

Chemical Identifiers

CAS 865-47-4
Molecular Formula C4H9KO
Molecular Weight (g/mol) 112.21
MDL Number MFCD00012162
InChI Key LPNYRYFBWFDTMA-UHFFFAOYSA-N
Synonym potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
PubChem CID 23665647
IUPAC Name potassium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426121000
SDS View Documents
Thermo Scientific Chemicals
426121000
100 mL AcroSeal™ Glass Bottle
Each for $153.93
 
AC426128000
SDS View Documents
Thermo Scientific Chemicals
426128000
800 mL AcroSeal™ Glass bottle
Each for $693.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

865-47-4
112.21
LPNYRYFBWFDTMA-UHFFFAOYSA-N
23665647
CC(C)(C)[O-].[K+]
C4H9KO
MFCD00012162
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
potassium;2-methylpropan-2-olate

Specifications

865-47-4
C4H9KO
MFCD00012162
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok
CC(C)(C)[O-].[K+]
112.21
112.21
AcroSeal™ Glass Bottle
0.82
Potassium tert-butoxide, 1M solution in tert-butanol
100 mL
(CH3)3COK
12°C
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium;2-methylpropan-2-olate
23665647
0.90 to 1.10M
Colorless to amber solution
0.8200g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

GHS Signal Word: Danger

EINECSNumber : 212-740-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only