missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$233.68 - $1042.95
Specifications
Chemical Name or Material | Potassium tert-butoxide |
---|---|
CAS | 865-47-4,109-99-9 |
Color | Colorless to Orange-Yellow |
Physical Form | Solution |
Linear Formula | (CH3)3COK |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC371221000
|
Thermo Scientific Chemicals
371221000 |
100 mL | AcroSeal™ Glass bottle |
Each for $233.68
|
|
||||
AC371220010
|
Thermo Scientific Chemicals
371220010 |
1 L | AcroSeal™ Glass Bottle |
Each for $1,042.95
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium tert-butoxide | |
Colorless to Orange-Yellow | |
(CH3)3COK | |
−19°C | |
LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
potassium;2-methylpropan-2-olate | |
112.21 | |
0.95 to 1.10M | |
0.9020g/mL |
865-47-4,109-99-9 | |
Solution | |
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289 | |
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
CC(C)(C)[O-].[K+] | |
23665647 | |
Pure | |
0.902 |
RUO – Research Use Only