missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium tetraborate tetrahydrate, 99.5%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 259570010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium tetraborate tetrahydrate | |
White | |
Powder | |
99.5% | |
K2B4O7·4H2O | |
Potassium biborate tetrahydrate | |
305.5 | |
99.5% | |
Plastic bottle |
850.0°C | |
12045-78-2 | |
1 kg | |
B4K2O7·4H2O | |
14,768 | |
Solubility in water: soluble. | |
305.5 | |
Extra Pure |
Chemical Identifiers
12045-78-2 | |
B4K2O7·4H2O | |
305.5 | |
Potassium biborate tetrahydrate |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
GHS P Statement
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only