Learn More
Potassium tetrabromopalladate(II), Pd 20.0%, Thermo Scientific Chemicals
$193.58 - $672.03
Chemical Identifiers
CAS | 13826-93-2 |
---|---|
Molecular Formula | Br4H4K2Pd |
Molecular Weight (g/mol) | 508.26 |
MDL Number | MFCD00049659 |
InChI Key | PBPKCWAOLDCQKM-UHFFFAOYSA-N |
Synonym | potassium tetrabromopalladate ii, dipotassium tetrabromopalladium 2-, dipotassium tetrabromopalladate, dipotassium tetrabromopalladate 2-, palladate 2-, tetrabromo-, potassium 1:2, palladate 2-, tetrabromo-, dipotassium, sp-4-1, palladous potassium bromide, dipotassium tetrabromopalladiumdiuide, dipotassium ion tetrabromopalladiumdiuide |
PubChem CID | 61688 |
IUPAC Name | dipotassium;tetrabromopalladium(2-) |
SMILES | [K+].[K+].Br.Br.Br.Br.[Pd--] |
Description
Potassium tetrabromopalladate(II) Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13826-93-2 | |
508.26 | |
PBPKCWAOLDCQKM-UHFFFAOYSA-N | |
61688 | |
[K+].[K+].Br.Br.Br.Br.[Pd--] |
Br4H4K2Pd | |
MFCD00049659 | |
potassium tetrabromopalladate ii, dipotassium tetrabromopalladium 2-, dipotassium tetrabromopalladate, dipotassium tetrabromopalladate 2-, palladate 2-, tetrabromo-, potassium 1:2, palladate 2-, tetrabromo-, dipotassium, sp-4-1, palladous potassium bromide, dipotassium tetrabromopalladiumdiuide, dipotassium ion tetrabromopalladiumdiuide | |
dipotassium;tetrabromopalladium(2-) |
Specifications
Red to Brown | |
Crystalline | |
Br4H4K2Pd | |
potassium tetrabromopalladate ii, dipotassium tetrabromopalladium 2-, dipotassium tetrabromopalladate, dipotassium tetrabromopalladate 2-, palladate 2-, tetrabromo-, potassium 1:2, palladate 2-, tetrabromo-, dipotassium, sp-4-1, palladous potassium bromide, dipotassium tetrabromopalladiumdiuide, dipotassium ion tetrabromopalladiumdiuide | |
PBPKCWAOLDCQKM-UHFFFAOYSA-N | |
dipotassium;tetrabromopalladium(2-) | |
61688 | |
Pd 20.0% | |
Hygroscopic |
13826-93-2 | |
1 g | |
MFCD00049659 | |
Soluble in water. | |
[K+].[K+].Br.Br.Br.Br.[Pd--] | |
508.26 | |
504.21 | |
Odorless | |
Potassium tetrabromopalladate(II) |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
EINECSNumber : 237-535-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only