Learn More
Potassium tetrachloroaurate(III) hydrate, 99%, Thermo Scientific Chemicals
$247.16 - $1239.59
Chemical Identifiers
CAS | 27988-75-6 |
---|---|
Molecular Formula | AuCl4K |
Molecular Weight (g/mol) | 377.86 |
MDL Number | MFCD00043088 |
InChI Key | IKHCGJWURGSQRT-UHFFFAOYSA-J |
Synonym | potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion |
PubChem CID | 91886546 |
IUPAC Name | potassium;tetrachlorogold(1-);hydrate |
SMILES | [K+].Cl[Au+3](Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2329203
|
Thermo Scientific Chemicals
B2329203 |
1 g |
Each for $247.16
|
|
|||||
AAB2329206
|
Thermo Scientific Chemicals
B2329206 |
5 g |
Each for $1,239.59
|
|
|||||
Description
Potassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium tetrachloroaurate(III) hydrate is used as primary and secondary intermediates. A Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.
Solubility
Soluble in water, ethanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
27988-75-6 | |
377.86 | |
IKHCGJWURGSQRT-UHFFFAOYSA-J | |
91886546 | |
[K+].Cl[Au+3](Cl)(Cl)Cl |
AuCl4K | |
MFCD00043088 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
potassium;tetrachlorogold(1-);hydrate |
Specifications
357°C (decomposition) | |
1 g | |
KAuCl4·xH2O | |
UN3260 | |
potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion | |
IKHCGJWURGSQRT-UHFFFAOYSA-J | |
potassium;tetrachlorogold(1-);hydrate | |
91886546 | |
99% |
27988-75-6 | |
AuCl4K | |
MFCD00043088 | |
14,7681 | |
Soluble in water,ethanol. | |
[K+].Cl[Au+3](Cl)(Cl)Cl | |
377.86 | |
377.88 (Anhydrous) | |
Potassium tetrachloroaurate(III) hydrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 237-190-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only