missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetrachloroplatinate(II), 46-47% Pt, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 195360250

 View more versions of this product

Catalog No. AC195360250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium tetrachloroplatinate(II)
10025-99-7
Crystalline Powder and/or Crystals
Cl4H4K2Pt
MFCD00011378
15, 7801
Solubility in water: 10g/L (20°C)
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
419.11
415.1
Glass bottle
250.0°C
Red to Brown
25 g
K2PtCl4
02,182; 04,215
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
JCAXKWJYKUEOFO-UHFFFAOYSA-N
dipotassium;tetrachloroplatinum(2-)
61440
46 to 47% Pt

Chemical Identifiers

10025-99-7
419.11
JCAXKWJYKUEOFO-UHFFFAOYSA-N
61440
Cl4H4K2Pt
MFCD00011378
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-050-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only