missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1560530

 View more versions of this product

Catalog No. AAA1560530


Explore available promotions

Description

Description

Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrafluoroborate is used as an active filler in the preparation of resin-bonded abrasives. It is also used for extraction, refining and processing of metals in the chemical industry. It finds application as additive in alloying industry and welding agent. Further, it is used in the production of fluxing agents for brazing and soldering.

Solubility
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Specifications

Specifications

(decomposition)
529°C
250 g
KBF4
14,7686
Slightly soluble in water and hot ethyl alcohol. Insoluble in cold ethyl alcohol.
[K+].F[B-](F)(F)F
125.90
125.91
1.3245
2.5
Potassium tetrafluoroborate
14075-53-7
BF4K
MFCD00011395
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
AKEBROIVCDHVSD-UHFFFAOYSA-N
potassium tetrafluoroboranuide
518872
98%
Hygroscopic

Chemical Identifiers

14075-53-7
125.90
AKEBROIVCDHVSD-UHFFFAOYSA-N
518872
[K+].F[B-](F)(F)F
BF4K
MFCD00011395
potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
potassium tetrafluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

EINECSNumber : 237-928-2

RTECSNumber : ED2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only