missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thioacetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0540522

 View more versions of this product

Catalog No. AAL0540522


Explore available promotions

Description

Description

Potassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thioacetate is used for palladium mediated coupling with aryl halides and triflates leading to S-arylthioacetates and derivatives. It is also used as a reagent in the conversion of halides to thiols.

Solubility
Soluble in water, methanol and some polar solvents. Insoluble in alkanes, ethers and most nonpolar solvents.

Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Specifications

Specifications

Potassium thioacetate
∽240°C (decomposition)
C2H4KOS
100 g
3595448
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
SDJHDRMYZQFJJO-UHFFFAOYSA-N
ethanethioic S-acid;potassium
24201352
98%
10387-40-3
Stench
MFCD00083065
UN3335
Air sensitive; Hygroscopic
Soluble in water,methanol and some polar solvents. Insoluble in alkanes,ethers and most nonpolar solvents.
CC(=O)S.[K]
115.211
114.21

Chemical Identifiers

10387-40-3
115.211
SDJHDRMYZQFJJO-UHFFFAOYSA-N
24201352
CC(=O)S.[K]
C2H4KOS
MFCD00083065
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
ethanethioic S-acid;potassium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

EINECSNumber : 233-848-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only