missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thioacetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 221300250

 View more versions of this product

Catalog No. AC221300250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium thioacetate
10387-40-3
≤11 (10% aq. soln.)
25 g
C2H3KOS
02,23
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
SDJHDRMYZQFJJO-UHFFFAOYSA-N
ethanethioic S-acid;potassium
24201352
98%
Glass bottle
173.0°C to 176.0°C
White to Brown
Crystalline Powder, Crystals and/or Chunks
98%
CH3COSK
01,955
Solubility in water: freely soluble in water
CC(=O)S.[K]
114.2
114.2
Authentic
(10% in water) clear

Chemical Identifiers

10387-40-3
114.2
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
ethanethioic S-acid;potassium
C2H3KOS
SDJHDRMYZQFJJO-UHFFFAOYSA-N
24201352
CC(=O)S.[K]
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 233-848-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only