missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium thioacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221301000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium thioacetate | |
10387-40-3 | |
≤11 (10% aq. soln.) | |
100 g | |
C2H3KOS | |
02,23 | |
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
ethanethioic S-acid;potassium | |
24201352 | |
98% | |
Glass bottle |
173.0°C to 176.0°C | |
White to Brown | |
Crystalline Powder, Crystals and/or Chunks | |
98% | |
CH3COSK | |
01,955 | |
Solubility in water: freely soluble in water | |
CC(=O)S.[K] | |
114.2 | |
114.2 | |
Authentic | |
(10% in water) clear |
Chemical Identifiers
10387-40-3 | |
114.2 | |
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium | |
ethanethioic S-acid;potassium |
C2H3KOS | |
SDJHDRMYZQFJJO-UHFFFAOYSA-N | |
24201352 | |
CC(=O)S.[K] |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 233-848-7
RUO – Research Use Only