missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thioacetate, 98%, Thermo Scientific Chemicals

$94.78 - $279.31

Chemical Identifiers

CAS 10387-40-3
Molecular Formula C2H3KOS
Molecular Weight (g/mol) 114.2
InChI Key SDJHDRMYZQFJJO-UHFFFAOYSA-N
Synonym potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
PubChem CID 24201352
IUPAC Name ethanethioic S-acid;potassium
SMILES CC(=O)S.[K]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221300250
SDS View Documents
Thermo Scientific Chemicals
221300250
25 g Glass bottle
Each for $94.78
 
AC221301000
SDS View Documents
Thermo Scientific Chemicals
221301000
100 g Glass bottle
Each for $279.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10387-40-3
114.2
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
ethanethioic S-acid;potassium
C2H3KOS
SDJHDRMYZQFJJO-UHFFFAOYSA-N
24201352
CC(=O)S.[K]

Specifications

173.0°C to 176.0°C
White to Brown
Crystalline Powder, Crystals and/or Chunks
98%
CH3COSK
01,955
Solubility in water: freely soluble in water
CC(=O)S.[K]
114.2
114.2
Authentic
(10% in water) clear
10387-40-3
≤11 (10% aq. soln.)
25 g
C2H3KOS
02,23
potassium thioacetate, ethanethioic s-acid; potassium, schiff reagent potassium
SDJHDRMYZQFJJO-UHFFFAOYSA-N
ethanethioic S-acid;potassium
24201352
98%
Glass bottle
Potassium thioacetate
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 233-848-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only