Learn More
Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035602K2
Description
Potassium thiocyanate, 0.1N standardized solution is used in analytical chemistry for the quantitative determination of Fe(III). In Volhardâs method, it is used to determine the concentration of chloride ion by titration. It is also used as an analyzer for direct and continuous measurement of the concentration of soluble copper in an aqueous solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium thiocyanate | |
333-20-0 | |
1 L | |
MFCD00011413 | |
Miscible with water,acetone and alcohol. | |
C(#N)[S-].[K+] | |
97.176 | |
CHEBI:30951 | |
Odorless | |
23 hPa (17mm Hg) at 20°C |
Colorless | |
Liquid | |
CKNS | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
potassium;thiocyanate | |
516872 | |
97.18 | |
Colorless liquid |
Chemical Identifiers
333-20-0 | |
97.176 | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
516872 | |
potassium;thiocyanate |
CKNS | |
MFCD00011413 | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
CHEBI:30951 | |
C(#N)[S-].[K+] |
Safety and Handling
EINECSNumber : 206-370-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only