missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 035602K7

 View more versions of this product

Catalog No. AA35602K7


Explore available promotions

Description

Description

Potassium thiocyanate, 0.1N standardized solution is used in analytical chemistry for the quantitative determination of Fe(III). In Volhard’s method, it is used to determine the concentration of chloride ion by titration. It is also used as an analyzer for direct and continuous measurement of the concentration of soluble copper in an aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium thiocyanate
333-20-0
4 L
MFCD00011413
Miscible with water,acetone and alcohol.
C(#N)[S-].[K+]
97.176
CHEBI:30951
Odorless
23 hPa (17mm Hg) at 20°C
Colorless
Liquid
CKNS
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
Colorless liquid

Chemical Identifiers

333-20-0
97.176
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
CKNS
MFCD00011413
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]
Safety and Handling

Safety and Handling

EINECSNumber : 206-370-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only