missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Trihydrogen Dioxalate Dihydrate 99.0+%, TCI America™
$44.67 - $257.73
Chemical Identifiers
CAS | 6100-20-5 |
---|---|
Molecular Formula | C4H3KO8 |
Molecular Weight (g/mol) | 218.16 |
MDL Number | MFCD00150443,MFCD00150443 |
InChI Key | GANDVAJEIJXBQJ-UHFFFAOYSA-M |
Synonym | potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate |
PubChem CID | 131698588 |
IUPAC Name | potassium oxalic acid hydrogen oxalate |
SMILES | [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O |
Chemical Identifiers
6100-20-5 | |
218.16 | |
GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
131698588 | |
[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O |
C4H3KO8 | |
MFCD00150443,MFCD00150443 | |
potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
potassium oxalic acid hydrogen oxalate |
Specifications
6100-20-5 | |
C4H3KO8 | |
25 g | |
potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate | |
[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O | |
218.16 | |
218.16 | |
Crystalline Powder |
White | |
MFCD00150443,MFCD00150443 | |
2811 | |
GANDVAJEIJXBQJ-UHFFFAOYSA-M | |
potassium oxalic acid hydrogen oxalate | |
131698588 | |
≥99.0% (T) | |
Potassium Trihydrogen Dioxalate Dihydrate |
Safety and Handling
EINECSNumber : (2)-0922
TSCA : Yes