missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium trioxalatoferrate(III) trihydrate, Thermo Scientific Chemicals

$73.78 - $1428.07

Chemical Identifiers

CAS 5936-11-8
Molecular Formula C6H3FeK3O12
Molecular Weight (g/mol) 440.22
MDL Number MFCD00150450
InChI Key YXTWCYYDGRBUDE-UHFFFAOYSA-K
Synonym Potassium iron(III) oxalate
PubChem CID 131675837
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3112406
View Documents
Thermo Scientific Chemicals
03112406
5 g
Each for $73.78
 
AA3112414
View Documents
Thermo Scientific Chemicals
03112414
25 g
Each for $144.81
 
AA3112422
View Documents
Thermo Scientific Chemicals
03112422
100 g
Each for $425.49
 
AA3112436
View Documents
Thermo Scientific Chemicals
03112436
500 g
Each for $1,428.07
 
Description

Description

Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5936-11-8
440.22
YXTWCYYDGRBUDE-UHFFFAOYSA-K
131675837
C6H3FeK3O12
MFCD00150450
Potassium iron(III) oxalate

Specifications

5936-11-8
Crystalline
C6H3FeK3O12
UN3288
Slightly soluble in water.
C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
440.22
491.26 (437.21 Anhydrous)
2.133 g/mL
Green
5 g
MFCD00150450
Potassium iron(III) oxalate
YXTWCYYDGRBUDE-UHFFFAOYSA-K
tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
131675837
Hygroscopic
Potassium trioxalatoferrate(III) trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 238-954-7

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only