missing translation for 'onlineSavingsMsg'
Learn More

Povidone-Iodine Solution USP, 10% (w/v), 1% (w/v) Available Iodine, for Laboratory Use Only, Ricca Chemical

For Laboratory Use

Supplier:  Ricca Chemical Company 395516

Catalog No. 3955-16

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Povidone-Iodine Solution
1% (w/v) Available Iodine
90.0,7.93
(C6H9NO)nI2
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex
II.*-CC(-*)N1CCCC1=O
364.95
10%
Amber Poly Bottle
500 mL
10% (w/v)
Brown
25655-41-8,7732-18-5
94.0,8.25
MFCD00084483
CPKVUHPKYQGHMW-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one; diiodine
410087
USP
1.5 to 4.4
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions