Eisco™ Plastic Powder Funnels
Eisco™ Plastic Powder Funnels are an ideal method for transferring powder reagents from one vessel to another.
$5.62 - $11.26
Specifications
Material | Polypropylene |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Transferring powdered reagents |
Includes | Parallel stem |
Product Type | Filling Funnel (Powders) |
Description
- Polypropylene powder funnel
- Designed for transferring powder reagents
- Translucent
- Durable for student use
- Easy to clean
- Autoclavable
- Chemical, acid, and alkali resistant
Specifications
Polypropylene | |
Transferring powdered reagents | |
Filling Funnel (Powders) |
Autoclavable | |
Parallel stem |