missing translation for 'onlineSavingsMsg'
Learn More

Alconox™ Powdered Precision Cleaner

Concentrated, anionic detergent for manual and ultrasonic cleaning

$1824.92 - $2089.43

Specifications

Product Type Powdered Precision Cleaner
Form Powder
pH 9.5
Appearance White, Cream
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
View More Specs

Products 4
Catalog Number Mfr. No. Volume (English) Container Type Price Quantity  
Catalog Number Mfr. No. Volume (English) Container Type Price Quantity  
04-322-5C
View Documents
Alconox™
1150
50 lb. Carton
Each for $2,089.43
Only null left
 
04-322-5D
View Documents
Alconox™
ALC1101
100 lb. Drum
Each for $1,824.92
Only null left
 
04-322-5B
View Documents
Alconox™
1125
25 lb. Carton
Each for $1,824.92
Only null left
 
16-000-108
View Documents
Alconox™
1103
300 lb. Drum
Each for $1,824.92
Only null left
 
Description

Description

Concentrated, anionic detergent for manual and ultrasonic cleaning

Alconox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, carbonates.

  • Biodegradable and readily disposable
  • Penetrating wetting power to save time
  • Replaces corrosive acids and hazardous solvents
  • Free rinsing gives reliable results without interfering residues
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • Dilute 1:100 for typical use (10g in 1L of water)
  • USDA authorized for use in federally inspected meat and poultry plants

Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, pipes, radioactive contaminated articles, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, tanks and reactors, passes inhibitory residue test for water analysis.

Used to remove:
Soil, grit, grime, buffing compound, slime, grease, oils, blood, tissue, salts, deposits, particulates, solvents, chemicals, radioisotopes, radioactive contaminations, silicon oils, mold release agents.

Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal,stainless steel, porcelain, ceramic, plastic, rubber and fiberglass, can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.

Shelf life 2 years from date of manufacture.

Order Info

0.5kg (1 lb.) makes 49.2L (13 gal.) cleaning solution.

Specifications

Specifications

Powdered Precision Cleaner
9.5
Store closed and upright in a cool dry place at 15° to 30°C.
Powder
White, Cream
Cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber and fiberglass
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'