missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Treston™ Power Rail with Upright Mounting Bracket
Mounting positions include horizontal and vertical on uprights, rear case mounting, front rail mounting, and work surface mounting. Power rails are available with surge protection.
Supplier: Treston™ 1491118810
Description
- 20 amp, 125 volt, NEMA 5-20P plug
- Black aluminum housing with on/off switch
- Upright mounting brackets included for horizontal or vertical mounting positions
Specifications
30.5 in. | |
Power Rail | |
51 mm | |
1 Ea. | |
30.5 in. | |
775 mm | |
775 mm |
M36 Power Rail | |
2 in. | |
2 in. | |
8 Outlets, Surge | |
51 mm | |
Treston Workstations | |
Steel |