missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ PowerTrack SYBR Green Master Mix with PCR Plates Bundle DFS Item

Switch to the affordable solution that eases optimization and pipetting efforts while delivering unsurpassed specificity. Choose the fast 0.1 mL PowerTrack SYBR Green Master Mix and Optical MicroAmp Reaction Plates and Adhesive covers bundle option for faster PCR.

$1044.72 - $1044.72

Specifications

Detection Method SYBR
Includes Label or Dye Yes
For Use With (Application) Real Time PCR (qPCR)
View More Specs

Products 1
Catalog Number Mfr. No. Includes Reaction Speed Price Quantity  
Catalog Number Mfr. No. Includes Reaction Speed Price Quantity  
A48344 DFS Item
View Documents Promotion Details
Applied Biosystems™
A48344
MicroAmp Fast Optical 96-well Reaction Plates, 0.1 mL Fast
Each for $1,044.72
 
Description

Description

Fast Bundle Includes (A48344)

Includes:

  • A46110 PowerTrack SYBR Green Master Mix 2 x 5 mL
  • 4483485 MicroAmp Fast Plates - 20 Plates
  • 4311971 MicroAmp Plate Covers - 100 Covers

Features:

  • Reduce total reaction time from 2 hours to 25 minutes
  • Two-color tracking dye reduces pipetting errors
  • Broad dynamic range eases optimization efforts
  • High specificity yields confident results
  • Combine with SuperScript IV VILO Master Mix for clearly better PCR

 

Standard Bundle Includes (A48345)

Includes:

  • A46110 PowerTrack SYBR Green Master Mix 2 x 5 mL
  • 4306737 MicroAmp Plates - 20 Plates
  • 4311971 MicroAmp Plate Covers - 100 Covers

Features:

  • MicroAmp PCR plates deliver unmatched uniformity
  • Two-color tracking dye reduces pipetting errors
  • Broad dynamic range eases optimization efforts
  • High sensitivity yields confident results
  • Combine with SuperScript IV VILO Master Mix for clearly better PCR
Specifications

Specifications

SYBR
Yes
Real Time PCR (qPCR)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'