Learn More
Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO), Thermo Scientific Chemicals
$590.18 - $1887.73
Chemical Identifiers
CAS | 7440-10-0 |
---|---|
Molecular Formula | Pr |
Molecular Weight (g/mol) | 140.91 |
MDL Number | MFCD00011174 |
InChI Key | PUDIUYLPXJFUGB-UHFFFAOYSA-N |
Synonym | unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg |
PubChem CID | 23942 |
ChEBI | CHEBI:49828 |
IUPAC Name | praseodymium |
SMILES | [Pr] |
Description
Praseodymium optical properties for use in amplification of telecommunication systems, including as a doping agent in fluoride fibers. It is also used in the scintillator for medical CAT scans. Used as additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Specifications
7440-10-0 | |
MFCD00011174 | |
25 x 25 mm | |
Reacts with water. | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
Odorless | |
≈0.42g/25x25mm | |
Praseodymium foil, 0.1mm (0.004 in.) thick |
Pr | |
UN3178 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
99.9% | |
Mylar under Argon | |
Foil | |
(REO) |
Safety and Handling
GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 231-120-3
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only