missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 00607FI
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Gray | |
0.62mm (0.024 in.) thick | |
(REO) | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
Odorless | |
≈2.6g/25x25mm |
Praseodymium foil | |
7440-10-0 | |
Pr | |
50 x 50 mm | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
99.5% | |
Mylar under Argon | |
Foil |
Chemical Identifiers
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Safety and Handling
EINECSNumber : 231-120-3
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only