missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO), Thermo Scientific Chemicals

$352.45 - $1190.67

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13963FF
View Documents
Thermo Scientific Chemicals
013963FF
25 x 25 mm
Each for $352.45
 
AA13963FI
View Documents
Thermo Scientific Chemicals
013963FI
50 x 50 mm
Each for $1,190.67
 
Description

Description

Praseodymium is used as an alloying constituent for alloys used to make permanent magnets and flints. Along with neodymium, praseodymium is used to manufacture yellow glass which can be used as eye protection (e.g. for welders).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-10-0
140.91
PUDIUYLPXJFUGB-UHFFFAOYSA-N
23942
praseodymium
Pr
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
CHEBI:49828
[Pr]

Specifications

White
Pr
25 x 25 mm
Insoluble in water.
[Pr]
140.91
CHEBI:49828
Odorless
≈4.23g/25x25mm
Praseodymium foil, 1.0mm (0.04 in.) thick
7440-10-0
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
99.5%
Mylar under Argon
Foil
(REO)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-120-3

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only