missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Praseodymium ingot, 99% (REO), Thermo Scientific Chemicals
$186.63 - $742.95
Chemical Identifiers
CAS | 7440-10-0 |
---|---|
Molecular Formula | Pr |
Molecular Weight (g/mol) | 140.91 |
MDL Number | MFCD00011174 |
InChI Key | PUDIUYLPXJFUGB-UHFFFAOYSA-N |
Synonym | unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg |
PubChem CID | 23942 |
ChEBI | CHEBI:49828 |
IUPAC Name | praseodymium |
SMILES | [Pr] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-10-0 | |
140.91 | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
23942 | |
praseodymium |
Pr | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
CHEBI:49828 | |
[Pr] |
Specifications
Gray to Silver | |
Gray to silver ingot | |
Pr | |
10 g | |
PUDIUYLPXJFUGB-UHFFFAOYSA-N | |
praseodymium | |
23942 | |
Odorless | |
Praseodymium, ingot |
7440-10-0 | |
99.9% | |
MFCD00011174 | |
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg | |
[Pr] | |
140.91 | |
CHEBI:49828 | |
Air and moisture sensitive |
Safety and Handling
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260-H500
DOTInformation : DOT Class: 4.1, Packaging Group: II
EINECSNumber : 231-120-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only