missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Praseodymium powder, -40 mesh, 99.5% (REO), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 04418306

Numéro de catalogue. AA4418306


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Spécifications

Spécifications

Gray to Silver
7440-10-0
Pr
UN3089
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
[Pr]
140.91
CHEBI:49828
−40 Mesh
Ampule
Powder
Praseodymium powder
(REO)
MFCD00011174
5 g
PUDIUYLPXJFUGB-UHFFFAOYSA-N
praseodymium
23942
99.9%
Odorless
Air and moisture sensitive

Identifiants chimiques

7440-10-0
140.91
PUDIUYLPXJFUGB-UHFFFAOYSA-N
23942
praseodymium
Pr
MFCD00011174
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
CHEBI:49828
[Pr]
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228-H500

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 231-120-3

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

FDS
Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only