missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pravastatin Sodium 98.0+%, TCI America™
$527.52 - $1776.44
Chemical Identifiers
| CAS | 81131-70-6 |
|---|---|
| Molecular Formula | C23H35NaO7 |
| Molecular Weight (g/mol) | 446.52 |
| MDL Number | MFCD00887601,MFCD00887601 |
| InChI Key | VWBQYTRBTXKKOG-IYNICTALSA-M |
| Synonym | pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol |
| PubChem CID | 16759173 |
| ChEBI | CHEBI:8361 |
| IUPAC Name | sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| SMILES | [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1796100MG
|
TCI America
P1796100MG |
100 mg |
Each for $527.52
|
|
|||||
|
P1796500MG
|
TCI America
P1796500MG |
500 mg |
Each for $1,776.44
|
|
|||||
Chemical Identifiers
| 81131-70-6 | |
| 446.52 | |
| VWBQYTRBTXKKOG-IYNICTALSA-M | |
| 16759173 | |
| sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| C23H35NaO7 | |
| MFCD00887601,MFCD00887601 | |
| pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol | |
| CHEBI:8361 | |
| [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 |
Specifications
| 173°C | |
| 81131-70-6 | |
| 100 mg | |
| MFCD00887601,MFCD00887601 | |
| VWBQYTRBTXKKOG-IYNICTALSA-M | |
| sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate | |
| 16759173 | |
| 446.52 | |
| Pravastatin Sodium |
| White-Yellow | |
| Crystalline Powder | |
| C23H35NaO7 | |
| pravastatin sodium, mevalotin, elisor, pravaselect, lipostat, selektine, pravachol, bristacol, prareduct, pravacol | |
| [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC([O-])=O)[C@@H]12 | |
| 446.52 | |
| CHEBI:8361 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
RTECSNumber : QJ7185000
TSCA : No
Recommended Storage : Refrigerator