missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GVS™ Pre-Sep™ Glass Fiber Membranes Without Binders
Glass prefilters without binders are designed to avoid binder extractables during solvent filtration or gravimetric analysis
Supplier: GVS™ 1215589
Description
- Composed of borosilicate glass fibers woven into a porous matrix without bonding resin
- Biologically inert
- Highly resistant to oxidizing agents and weak acids
- Filters without binders are recommended for analytical and gravimetric determinations.
Specifications
Autoclavable | |
Glass Fiber | |
Sterile | |
Glass fiber filter binderless |
White | |
1.0 μm | |
500 | |
47 mm |