missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Precast Gel Plus, Tris-Gly 15%
These precast polyacrylamide gels are suitable for both SDS-PAGE and Native-PAGE electrophoresis followed by gel staining or Western blotting.
$188.00 - $188.00
Specifications
Content And Storage | 4°C for 12 Months |
---|---|
Buffer | MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE |
Color | Clear |
Concentration | 15% |
For Use With (Equipment) | Bio-Rad Mini Protean Tanks |
Catalog Number | Mfr. No. | Sample Loading Volume | Wells | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Loading Volume | Wells | Price | Quantity | ||||
ICN08W00005
|
MP Biomedicals™
08W00005 |
≤50 μL | 10-well |
Pack of 10 Each for $188.00
|
|
||||
ICN08W00006
|
MP Biomedicals™
08W00006 |
≤30 μL | 15-well |
Pack of 10 Each for $188.00
|
|
||||
Description
Precast Gel Plus gels are an efficient and convenient scientific tool for separating peptides and small proteins with clear band resolution. In addition, they are compatible with Bio-Rad Mini Protean Tanks, thus facilitating the use of our gels without the need to replace your current hardware.
- Ready-to-use gels for SDS-PAGE or Native-PAGE applications
- Resilient to mechanical stretching during handling
- Removable comb prevents well-wall breakage
- Outlined and numbered wells facilitate gel loading and tracking
- Easy to store and stable at room temperature for 6 months
- Do not freeze
Specifications
4°C for 12 Months | |
Clear | |
Bio-Rad Mini Protean Tanks | |
Mini | |
Tris-Glycine | |
Peptides, Proteins | |
Denaturing, Native | |
10 Cassettes |
MP Bio™ Ultra-Fast Running Buffer for SDS-PAGE | |
15% | |
15% | |
1 mm | |
Precast Gel | |
10 to 100 kDa | |
Gel Staining SDS-PAGE, Native-PAGE, Blue Native, Western Blot, Acrylamide, Tris-Glycine, Mini Gels, Electrophoresis |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.