Learn More
Thermo Scientific™ Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
$4876.88 - $6848.37
Specifications
Controller Type | Digital |
---|---|
Certifications/Compliance | UL, CE |
Communications | Digital |
Temperature Range | Ambient +5° to 100°C |
Display | Monochrome LCD |
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Electrical Requirements | Model | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Capacity (Metric) | Electrical Requirements | Model | Price | Quantity | |||
TSCIR19
|
Thermo Scientific™
TSCIR19 |
19L Circulating Water Bath | 19 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 19 |
Each for $4,876.88
|
|
|||
TSCIR35
|
Thermo Scientific™
TSCIR35 |
35L Circulating Water Bath | 35 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 35 |
Each for $5,356.78
|
|
|||
TSCIR89
|
Thermo Scientific™
TSCIR89 |
89L Circulating Water Bath | 89 L | Global Voltage 100/115 V 200/230 V 50/60 Hz | Precision CIR 89 |
Each for $6,848.37
|
|
|||
Description
Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
- Achieve enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Optimize scheduling with auto-on and auto-off timers
- Accommodate taller labware with new hinged lid and extended height
- Help prevent bath overheating and damage with low-fluid protection
- Easily operate and monitor with icon-based graphical display
- Protect your work with audible alarms
Specifications
Digital | |
Digital | |
Monochrome LCD | |
304 Stainless Steel | |
±0.1°C | |
Global Voltage 100/115 V 200/230 V 50/60 Hz |
UL, CE | |
Ambient +5° to 100°C | |
1 Stainless Steel Gable cover and Rubber Duck | |
4 | |
±0.05°C |