Precision Laboratories Glucose Test Strips
Used in biological courses for detection of glucose concentrations.
Supplier: Precision Laboratories GLU1B100
Description
- Used in osmosis and diffusion experiments, where one is looking for differences in the diffusion of starch when compared to glucose, in a semi permeable membrane system
- Concentration ranges: 0, 100, 300, 1000 and 3000mg/dL
Specifications
Glucose | |
Urinalysis, used in biological courses for detection of Glucose concentrations | |
100 Pack | |
1 to 2 sec. |