missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Precoated HPTLC Glass Plates

Smaller size and distribution of HPTLC media yields improved resolution, shorter analysis times, and higher detection sensitivity

Supplier:  MilliporeSigma™ 1056310001

 View more versions of this product

Catalog No. M1056310001


Explore available promotions

Description

Description

  • Average particle size is 5μm with a 4-7μm distribution
  • Untreated, chemically modified, and reversed-phase silica gel with a pore size of 60Å

For all common gel separations, including weak to strong polar analytes, for applications requiring charring procedures

Certifications

ISO 9001:2000

Specifications

Specifications

25/Pk.
Silica Gel 60
10 x 10 cm
Silica HPTLC Plate
200 μm
SDS
Documents

Documents

Promotions

Promotions