missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Precoated HPTLC Glass Plates
Smaller size and distribution of HPTLC media yields improved resolution, shorter analysis times, and higher detection sensitivity
Supplier: MilliporeSigma™ 1056440001
Description
- Average particle size is 5μm with a 4-7μm distribution
- Untreated, chemically modified, and reversed-phase silica gel with a pore size of 60Å
For all common gel separations, including weak to strong polar analytes, for applications requiring charring procedures
Certifications
ISO 9001:2000
Specifications
100/Pk. | |
Silica Gel 60 Prescored | |
5 x 5 cm |
Silica HPTLC Plate | |
200 μm |