missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Precoated TLC and PLC Glass Plates
Average particle size is 15μm with a 5-20μm distribution
Supplier: MilliporeSigma™ 1117640001
Description
- Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
- 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
- Silica can also be used for applications requiring charring procedures
- Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
- For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates
Certifications
ISO 9001:2000
†With acid-stable fluorescent indicator.
‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.
Specifications
3 to 20 min. | |
ISO 9001:2000 | |
5 to 6 μm | |
25/Pk. | |
254 nm | |
36.0 | |
0.1 to 0.5 μL | |
250 μm | |
10 cm | |
0.0005 in. |
Glass | |
Silica Gel 60 F254 AMD, extra thin | |
60 Å | |
Silica HPTLC Plate | |
High Performance, Completely Contamination-Free Separations Especially in Demanding Pharmacopeial AppliCatIons | |
4 to 8 μm | |
Silica Gel 60 F254 AMD, extra thin | |
20 x 10 cm | |
10 cm | |
12 μm |