MilliporeSigma™ Precoated TLC and PLC Glass Plates
Average particle size is 15μm with a 5-20μm distribution
Supplier:  MilliporeSigma™ 1156850001
 View more versions of this product
Catalog No. M1156850001
Description
Description
  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates
Certifications
ISO 9001:2000
†With acid-stable fluorescent indicator.
‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.
Specifications
Specifications
Glass
Silica Gel 60 RP-8 F254 s
25/Pk.
254 nm (Acid Stable)
Silica Gel 60 RP-18 F254
20 x 20 cm
5 cm
ISO 9001:2000
60 Å
RP-modified Silica TLC Plate
Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems
250 μm
10 cm