Contec™ PREempt™ RTU Disinfectant Solution
PREempt™ RTU utilizes AHP™, a patented blend when combined with low levels of hydrogen peroxide, increase efficacy and cleaning performance. When combined, it creates an ideal one step cleaner/disinfectant.
Supplier:  Contec™ 11105
 View more versions of this product
Catalog No. 29-636-6213
Explore available promotions
Description
Description
  • AHP™ balances fast contact time while maintaining cleaning efficacy
    • Carries no hazard classification
  • PREempt™ RTU claims:
    • Broad-spectrum sanitizing claim in 30 seconds
    • Bactericidal and general virucide and tuberculocidal 3 minute kill claim
  • Contains no volatile organic compounds
  • Leaves minimal residues on surfaces once dry
  • Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and lab areas (work stations, fume hoods, equipment, hard non-porous environmental surfaces)
  • Effective, fast-acting, one-step disinfectant cleaner
  • Convenient, ready-to-use disinfectant for daily use
  • Low residue and compatible with a wide range of materials
  • Odor-free for good operator acceptability
  • Does not require PPE
  • Hydrogen peroxide breaks down into water and oxygen for easy disposal and minimal, to no environmental impact
  • Not classified as dangerous goods
Specifications
Specifications
Disinfectant Cleaner
Non-sterile
4/Cs.
3.8 L