Contec™ PREempt™ RTU Disinfectant Wipes
PREempt™ RTU utilizes AHP™, a patented blend when combined with low levels of hydrogen peroxide, increase efficacy and cleaning performance. When combined, it creates an ideal one step cleaner/disinfectant.
Supplier:  Contec™ 11221
Catalog No. 29-636-6214
Explore available promotions
Description
Description
  • AHP™ has fast contact time while maintaining cleaning efficacy
  • Carries no hazard classification
  • Broad-spectrum sanitizing claim in 30 seconds as well as bactericidal, virucide, fungicidal and tuberculocidal 3 min kill claim
  • Contains no VOCs
  • Leaves minimal residues on surfaces once dry
  • Convenient presaturated wipes in pull top canister with containing 160 wipes/canister
  • All ingredients listed on EPA and Health Canada Inerts lists and FDA GRAS
  • Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and lab areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces
  • Effective, fast-acting, one-step disinfectant cleaner
  • Convenient meltblown polypropylene presaturated wipes
  • Low residue and compatible with a wide range of materials
  • Odor-free for good operator acceptability
  • Does not require PPE
  • Hydrogen peroxide breaks down into water and oxygen for easy disposal
    • Minimal to no environmental impact
  • Not classified as dangerous goods
Specifications
Specifications
Wipes
15.2 x 17.8 cm
160/Canister; 12/Case
Non-sterile
For cleaning and disinfecting labs, cleanrooms, work stations, fume hoods, equipment, hard non-porous environmental surfaces