missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Premium Glass Double Convex Lens

Round, premium glass double convex lens designed for physical science demonstrations and experiments

$7.76 - $17.79

Specifications

Material Glass
Diameter (Metric) 50 mm
For Use With (Application) Physical Science Demonstrations and Experiments
Product Type Convex Lens
View More Specs

Products 4
Catalog Number Mfr. No. Length (Metric) Focal Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Focal Diameter (Metric) Price Quantity  
S27702
View Documents
Eisco™
PH0531ACV
50 mm 50 mm
Each for $17.79
 
S27383
View Documents
Eisco™
PH0531BCV
100 mm 50 mm
Each for $10.71
 
S27703
View Documents
Eisco™
PH0531DCV
250 mm 50 mm
Each for $7.76
 
S27385
View Documents
Eisco™
PH0531CCV
150 mm 50 mm
Each for $7.82
 
Description

Description

  • High optical clarity
  • For educational use only
Specifications

Specifications

Glass
50 mm
Physical Science Demonstrations and Experiments
Convex Lens
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'