Air-Tite™ Premium Hypodermic Needles for Lab/Vet Use
Sterile, disposable needles for laboratory and veterinary use
Supplier:  Air-Tite™ N163D
 View more versions of this product
Catalog No. 14-817-103
Explore available promotions
Description
Description
Ideal for laboratory and veterinary use; sterile and disposable.
  • Stainless-steel
  • Regular-wall design needles; all have 12° regular bevel and tri-bevel points
  • High-density polypropylene hub fits any Luer-Lok™ or Luer-Slip syringe tip
  • Come in strips of individual blister packs
Specifications
Specifications
HDPP (Hub), Stainless Steel
0.065 in.
3 in.
12° Regular Bevel and Tri-bevel Points
Sterile
16 G
1.66 mm
Hypodermic Needle
Luer Lock, Luer Slip