missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Prepared Bottled Media: Tryptic Soy Broth, Wide Mouth Bottle
For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples
$179.53 - $323.89
Specifications
Cap Type | Lug cap |
---|---|
Color | Amber |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Tryptic Soy Broth |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
22-349-999
|
BD
257488 |
90 mL |
Pack of 10 Each for $323.89
|
|
||||||
22-349-998
|
BD
257486 |
100 mL |
Pack of 10 Each for $179.53
|
|
||||||
Description
- Ready for immediate use
- Sterility Assurance Level of 10-6 reduces risk of false positives
- Terminally autoclaved
- Conforms to Harmonized USP, EP, and JP requirements for sterility tests and to CLSI requirements for recovery
Specifications
Lug cap | |
Liquid | |
Tryptic Soy Broth | |
130 mm |
Amber | |
United States | |
Bottle | |
48 mm |