Prepared Individual Microscope Slides
High-quality microscope slides, manufactured under rigorous control to meet all scientific requirements.
Supplier:  Customer Channels Group (Ccg) MA111C
 View more versions of this product
Catalog No. S99460
Explore available promotions
Description
Description
  • Premium glass with fine ground edges
  • Size: 1 × 3 in. (26 × 76mm)
  • Specially stained to ensure a clear, multicolored representation of all tissue structures
  • Specimens are characteristic and representative of the taxonomic group and/or subject
  • All slides sold separately
Specifications
Specifications
Squamous epithelium, isolated cells from human mouth, smear
Prepared Slide