missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Prepared Microscope Slide - Hyaline Cartilage Section From Mammal
Prepared microscope slide of mammalian hyaline cartilage
Supplier: Eisco™ BS18045
Description
- Pre-cleaned glass slide with ground edges
- Professionally stained samples for best visualization
- Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
- Shows characteristic histology of hyaline cartilage
- Tissue section
Specifications
1 | |
Microbiology Slide | |
China | |
Cardboard Mailer | |
Rat | |
Prepared Slide |
10% | |
Microscopy | |
Formalin | |
1 | |
1 x 3 in. |