BD Presence-Absence Broth Water Testing Media
Determines the presence of pollution-indicating organisms in treated or drinking water
Supplier:  BD 219200
Catalog No. DF0019-17-0
Explore available promotions
Description
Description
  • Color changes from purple to yellow if lactose-fermenting organisms are present
  • Gas production is demonstrated by gently shaking the bottle
  • 175/8 oz. (500g)
Specifications
Specifications
Beef extract 3g, Peptone 5g, Lactose 7.46g, Pancreatic digest of Casein 5.9g, Proteose Peptone 3.93g, Dipotassium Phosphate 1.35g, Monopotassium Phosphate 1.35g, Sodium Chloride 2.46g, Sodium Lauryl Sulfate 0.05g, Bromcresol Purple 8.5mg
Determines the presence of pollution-indicating organisms in treated or drinking water
17.6 oz.